missing translation for 'onlineSavingsMsg'
Learn More

3,5-Di-tert-butylbenzoic acid, 99%

Product Code. 11995001 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11995001 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11995001 Supplier Thermo Scientific Acros Supplier No. 438220050
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 16225-26-6 | C15H22O2 | 234.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 16225-26-6
CAS Min % 98.5
CAS Max % 100.0
Melting Point 170.0°C to 173.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Molecular Formula C15H22O2
Quantity 5 g
Synonym 3,5-di-tert-butylbenzoic acid, 3,5-di-t-butylbenzoic acid, 3,5-ditertbutylbenzoic acid, 3,5-di-tert-butylbenzoicacid, 3,5-bis tert-butyl benzoic acid, benzoic acid, 3,5-bis 1,1-dimethylethyl, pubchem4953, acmc-209do2, 3,5-di-tertbutylbenzoic acid, ksc494i2n
InChI Key NCTSLPBQVXUAHR-UHFFFAOYSA-N
SMILES CC(C)(C)C1=CC(=CC(=C1)C(=O)O)C(C)(C)C
IUPAC Name 3,5-ditert-butylbenzoic acid
Molecular Weight (g/mol) 234.34
PubChem CID 85339
Formula Weight 234.34
Percent Purity 99%
Physical Form Crystalline Powder or Powder
Chemical Name or Material 3, 5-Di-tert-butylbenzoic acid
Show More Show Less
compliance-icons
Product Identifier
  • 3,5-Di-tert-butylbenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.