missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichlorobenzonitrile, TRC

Product Code. 30388273
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. Quantity unitSize
30388273 1 g 1g
1 options
This item is not returnable. View return policy

Product Code. 30388273

Brand: Toronto Research Chemicals TRCD4365281G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3,5-Dichlorobenzonitrile
CAS 6575-00-4
Molecular Formula C7H3Cl2N
Quantity 1 g
Synonym NSC 109596
InChI Formula InChI=1S/C7H3Cl2N/c8-6-1-5(4-10)2-7(9)3-6/h1-3H
SMILES Clc1cc(Cl)cc(c1)C#N
Molecular Weight (g/mol) 172.01
Formula Weight 170.9643
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.