missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichlorobenzylamine, 94%

Product Code. 11308595
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11308595 5 g 5g
11398585 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11308595 Supplier Thermo Scientific Alfa Aesar Supplier No. L12054.06
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 39989-43-0 | C7H7Cl2N | 176.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39989-43-0
Molecular Formula C7H7Cl2N
Molecular Weight (g/mol) 176.04
MDL Number MFCD00052681
InChI Key ICIJWOWQUHHETJ-UHFFFAOYSA-N
Synonym 3,5-dichlorobenzylamine, 3,5-dichlorophenyl methanamine, benzenemethanamine, 3,5-dichloro, 3,5-dichloro-benzylamine, 1-3,5-dichlorophenyl methanamine, 3,5-dichlorophenyl methylamine, pubchem23431, 3.5-dichlorobenzylamine, acmc-1cthv, integrase inhibitor, r1 4
PubChem CID 457602
SMILES NCC1=CC(Cl)=CC(Cl)=C1

Specifications

Density 1.32
Boiling Point 141°C to 144°C (15 mmHg)
Refractive Index 1.576
Quantity 5 g
UN Number UN2735
Beilstein 3237459
Sensitivity Air Sensitive
IUPAC Name 1-(3,5-dichlorophenyl)methanamine
Formula Weight 176.05
Percent Purity 97%
Chemical Name or Material 3,5-Dichlorobenzylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.