missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethoxybenzoyl Chloride, TRC

Product Code. 30377894
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
This item is not returnable. View return policy

Product Code. 30377894

Brand: Toronto Research Chemicals TRCD472623500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3,5-Dimethoxybenzoyl Chloride
CAS 17213-57-9
Molecular Formula C9H9ClO3
Quantity 500 mg
InChI Formula InChI=1S/C9H9ClO3/c1-12-7-3-6(9(10)11)4-8(5-7)13-2/h3-5H,1-2H3
SMILES COc1cc(OC)cc(c1)C(=O)Cl
IUPAC Name 3,5-dimethoxybenzoyl chloride
Molecular Weight (g/mol) 200.62
Formula Weight 200.024
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.