missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethoxybenzyl alcohol, 99%

Product Code. 11486197
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11486197 5 g 5g
11476197 1 g 1g
11496197 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11486197 Supplier Thermo Scientific Alfa Aesar Supplier No. B25138.06
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 705-76-0 | C9H12O3 | 168.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 705-76-0
Molecular Formula C9H12O3
Molecular Weight (g/mol) 168.19
MDL Number MFCD00004641
InChI Key AUDBREYGQOXIFT-UHFFFAOYSA-N
Synonym 3,5-dimethoxybenzyl alcohol, 3,5-dimethoxyphenyl methanol, 3,5-dimethoxybenzylalcohol, benzenemethanol, 3,5-dimethoxy, 3,5-dimethoxy benzyl alcohol, 3,5-dimethoxyphenyl methan-1-ol, 3,5-dimethoxybenzyl alcohole, pubchem7732, 3,5-dimethoxy-benzylalcohol, 3,5-dimethoxy benzylalcohol
PubChem CID 69718
IUPAC Name (3,5-dimethoxyphenyl)methanol
SMILES COC1=CC(OC)=CC(CO)=C1

Specifications

Melting Point 46°C to 50°C
Boiling Point 183°C to 186°C (20 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
Beilstein 1866305
Formula Weight 168.19
Percent Purity 99%
Chemical Name or Material 3,5-Dimethoxybenzyl alcohol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.