missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethyl-1-phenyl-1H-pyrazole-4-carbonitrile, ≥97%, Thermo Scientific™

Product Code. 10470564
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10470564 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10470564 Supplier Thermo Scientific Maybridge Supplier No. CC48016DA
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 23198-55-2
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C12H11N3
MDL Number MFCD08435888
Quantity 1 g
Synonym 3,5-dimethyl-1-phenyl-1h-pyrazole-4-carbonitrile, 1h-pyrazole-4-carbonitrile,3,5-dimethyl-1-phenyl
InChI Key WOFAIVGOIVCZJJ-UHFFFAOYSA-N
SMILES CC1=C(C(=NN1C2=CC=CC=C2)C)C#N
IUPAC Name 3,5-dimethyl-1-phenylpyrazole-4-carbonitrile
Molecular Weight (g/mol) 197.241
PubChem CID 12208701
Formula Weight 197.24
Percent Purity 97%
Chemical Name or Material 3, 5-Dimethyl-1-phenyl-1H-pyrazole-4-carbonitrile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.