missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylbenzeneboronic acid, 98%

Product Code. 11485407
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11485407 25 g 25g
11465407 1 g 1g
11475407 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11485407 Supplier Thermo Scientific Alfa Aesar Supplier No. B23434.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 172975-69-8 | C8H11BO2 | 149.984 g/mol

3,5-Dimethylbenzeneboronic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dimethylbenzeneboronic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 172975-69-8
Molecular Formula C8H11BO2
Molecular Weight (g/mol) 149.984
MDL Number MFCD00185689
InChI Key DJGHSJBYKIQHIK-UHFFFAOYSA-N
Synonym 3,5-dimethylphenyl boronic acid, 3,5-dimethylbenzeneboronic acid, 3,5-dimethylphenyl boranediol, m-xylene-5-boronic acid, boronic acid, 3,5-dimethylphenyl, pubchem1833, acmc-209e5l, ksc174i6l, 3.5-dimethylphenylboronic acid
PubChem CID 2734349
IUPAC Name (3,5-dimethylphenyl)boronic acid
SMILES B(C1=CC(=CC(=C1)C)C)(O)O

Specifications

Melting Point 235°C to 237°C
Quantity 25 g
Beilstein 7368658
Formula Weight 149.99
Percent Purity 98%
Chemical Name or Material 3,5-Dimethylbenzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.