missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylphenylboronic acid, 97%, Thermo Scientific™

Product Code. 10520594 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10520594 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10520594 Supplier Thermo Scientific Acros Supplier No. 359290050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 172975-69-8
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C8H11BO2
Synonym 3,5-dimethylphenyl boronic acid, 3,5-dimethylbenzeneboronic acid, 3,5-dimethylphenyl boranediol, m-xylene-5-boronic acid, boronic acid, 3,5-dimethylphenyl, pubchem1833, acmc-209e5l, ksc174i6l, 3.5-dimethylphenylboronic acid
InChI Key DJGHSJBYKIQHIK-UHFFFAOYSA-N
SMILES B(C1=CC(=CC(=C1)C)C)(O)O
IUPAC Name (3,5-dimethylphenyl)boronic acid
Molecular Weight (g/mol) 149.984
PubChem CID 2734349
Formula Weight 149.98
Percent Purity ≥96.0%
Chemical Name or Material 3, 5-Dimethylphenylboronic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.