missing translation for 'onlineSavingsMsg'
Learn More

3,6-Dibromopyridazide, 97+%, Maybridge

Product Code. 10126892
Change view
Click to view available options
Amber Glass Bottle; 1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10126892 Amber Glass Bottle; 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10126892 Supplier Thermo Scientific Maybridge Supplier No. MO07136DA

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 17973-86-3
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C4H2Br2N2
Quantity 1 g
Synonym 3,6-dibromopyridazide, pyridazine, 3,6-dibromo, 3,6-dibromo pyridazine, pubchem12385, 3,6-dibromo-pyridazine, pyridazine,3,6-dibromo, 3,6-dibromo-1,2-diazine, 3,6-dibromopyridazine, synquest 3h32-9-t3
InChI Key VQAFMTSSCUETHA-UHFFFAOYSA-N
SMILES C1=CC(=NN=C1Br)Br
IUPAC Name 3,6-dibromopyridazine
Molecular Weight (g/mol) 237.882
PubChem CID 248852
Formula Weight 237.88
Percent Purity 97%
Chemical Name or Material 3, 6-Dibromopyridazine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.