Learn More
3,6-Dimethyl-1,4-dioxane-2,5-dione, 95%, BLD Pharm™
Specifications
Specifications
| CAS | 95-96-5 |
| Molecular Formula | C6H8O4 |
| MDL Number | MFCD00011685 |
| Quantity | 100 g |
| SMILES | O=C(C(C)O1)OC(C)C1=O |
| Molecular Weight (g/mol) | 144.13 |
| Percent Purity | 95% |
| Chemical Name or Material | 3,6-Dimethyl-1,4-dioxane-2,5-dione |