missing translation for 'onlineSavingsMsg'
Learn More

3,6-Dimethyl-fluorene, TRC

Product Code. 17913755
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17913755 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17913755 Supplier Toronto Research Chemicals Supplier No. TRCD446815100MG
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7495-37-6
Molecular Formula C15H14
Quantity 100 mg
Synonym 3,6-Dimethyl-9H-fluorene, NSC 407702
InChI Formula InChI=1S/C15H14/c1-10-3-5-12-9-13-6-4-11(2)8-15(13)14(12)7-10/h3-8H,9H2,1-2H3
SMILES Cc1ccc2Cc3ccc(C)cc3c2c1
Molecular Weight (g/mol) 194.27
Chemical Name or Material 3,6-Dimethyl-fluorene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.