Learn More
3,7-Dimethyloct-6-en-1-ol, 95%, BLD Pharm™
Specifications
Specifications
| CAS | 106-22-9 |
| Molecular Formula | C10H20O |
| MDL Number | MFCD00002935 |
| Quantity | 1 kg |
| SMILES | CC(CCC=C(C)C)CCO |
| Molecular Weight (g/mol) | 156.27 |
| Percent Purity | 95% |
| Chemical Name or Material | 3,7-Dimethyloct-6-en-1-ol |