missing translation for 'onlineSavingsMsg'
Learn More

3-Acetyl-11-keto-Beta-boswellic Acid, TRC

Product Code. 30138281
Change view
Click to view available options
Quantity:
1 mg
10 mg
100 mg
Unit Size:
100mg
10mg
1mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138281 100 mg 100mg
17962685 10 mg 10mg
17943085 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138281 Supplier Toronto Research Chemicals Supplier No. TRCA185015100MG
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67416-61-9
Molecular Formula C32 H48 O5
Molecular Weight (g/mol) 512.72
Synonym 3-O-Acetyl-11-keto-β-boswellic Acid
IUPAC Name (3R,4R,4aR,6aR,6bS,8aR,11R,12S,12aR,14aR,14bS)-3-acetyloxy-4,6a,6b,8a,11,12,14b-heptamethyl-14-oxo-1,2,3,4a,5,6,7,8,9,10,11,12,12a,14a-tetradecahydropicene-4-carboxylic acid
SMILES C[C@@H]1CC[C@]2(C)CC[C@]3(C)C(=CC(=O)[C@@H]4[C@@]5(C)CC[C@@H](OC(=O)C)[C@@](C)([C@@H]5CC[C@@]34C)C(=O)O)[C@@H]2[C@H]1C

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C32H48O5/c1-18-9-12-28(4)15-16-30(6)21(25(28)19(18)2)17-22(34)26-29(5)13-11-24(37-20(3)33)32(8,27(35)36)23(29)10-14-31(26,30)7/h17-19,23-26H,9-16H2,1-8H3,(H,35,36)/t18-,19+,23-,24-,25+,26-,28-,29+,30-,31-,32-/m1/s1
Formula Weight 512.3502
Percent Purity >95%
Chemical Name or Material 3-O-Acetyl-11-keto-beta-Boswellic acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.