missing translation for 'onlineSavingsMsg'
Learn More

3-[[[[[[[(Acetylamino)carbonyl]amino]carbonyl]amino]carbonyl]amino]sulfonyl]-2-thiophenecarboxylic Acid Methyl Ester (Technical Grade), TRC

Product Code. 30138058
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138058 10 mg 10mg
30006729 5 mg 5mg
30004682 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138058 Supplier Toronto Research Chemicals Supplier No. TRCA16820810MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 171628-01-6
Molecular Formula C11H12N4O8S2
Molecular Weight (g/mol) 392.36
Synonym Methyl 3-(N-(((acetylcarbamoyl)carbamoyl)carbamoyl)sulfamoyl)thiophene-2-carboxylate, Impurity of Thiophanate-methyl
IUPAC Name methyl 3-(acetylcarbamoylcarbamoylcarbamoylsulfamoyl)thiophene-2-carboxylate
SMILES COC(=O)c1sccc1S(=O)(=O)NC(=O)NC(=O)NC(=O)NC(=O)C

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C11H12N4O8S2/c1-5(16)12-9(18)13-10(19)14-11(20)15-25(21,22)6-3-4-24-7(6)8(17)23-2/h3-4H,1-2H3,(H4,12,13,14,15,16,18,19,20)
Formula Weight 392.0097
Chemical Name or Material 3-((((((((Acetylamino)carbonyl)amino)carbonyl)amino)carbonyl)amino)sulfonyl)-2-thiophenecarboxylic Acid Methyl Ester (Technical Grade)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.