missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-1-methyl-1H-indazole, 95%

Product Code. 10555680 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10555680 5 g 5g
10471050 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10555680 Supplier Thermo Scientific Acros Supplier No. 435720050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

CAS: 60301-20-4 | C8H9N3 | 147.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60301-20-4
Molecular Formula C8H9N3
Molecular Weight (g/mol) 147.18
MDL Number MFCD09054755
InChI Key NYLGITXFVVEBLZ-UHFFFAOYSA-N
Synonym 1-methyl-1h-indazol-3-amine, 3-amino-1-methylindazole, 3-amino-1-methyl-1h-indazole, 1h-indazol-3-amine, 1-methyl, n-methyl-3-aminoindazole, 1-methyl-1h-3-indazoleamine, 1-methyl-1h-indazole-3-amine, 1-methyl-1h-indazol-3-ylamine, 1-methyl-1h-indazole-3-ylamine
PubChem CID 12291317
SMILES CN1N=C(N)C2=CC=CC=C12

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 95°C to 99°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Quantity 5 g
IUPAC Name 1-methylindazol-3-amine
Formula Weight 147.18
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-1-methyl-1H-indazole
Show More Show Less
compliance-icons
Product Identifier
  • 3-Amino-1-methyl-1H-indazole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.