missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-1-methyl-1H-indazole, 97%

Product Code. 15467217
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15467217 1 g 1g
15477217 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15467217 Supplier Thermo Scientific Alfa Aesar Supplier No. H34285.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

CAS: 60301-20-4 | C8H9N3 | 147.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60301-20-4
Molecular Formula C8H9N3
Molecular Weight (g/mol) 147.18
MDL Number MFCD09054755
InChI Key NYLGITXFVVEBLZ-UHFFFAOYSA-N
Synonym 1-methyl-1h-indazol-3-amine, 3-amino-1-methylindazole, 3-amino-1-methyl-1h-indazole, 1h-indazol-3-amine, 1-methyl, n-methyl-3-aminoindazole, 1-methyl-1h-3-indazoleamine, 1-methyl-1h-indazole-3-amine, 1-methyl-1h-indazol-3-ylamine, 1-methyl-1h-indazole-3-ylamine
PubChem CID 12291317
SMILES CN1N=C(N)C2=CC=CC=C12

Specifications

Melting Point 95°C to 97°C
Quantity 1 g
IUPAC Name 1-methylindazol-3-amine
Formula Weight 147.18
Percent Purity 97%
Chemical Name or Material 3-Amino-1-methyl-1H-indazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.