missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-2,2-dimethyl-1-propanol, 97%, Thermo Scientific™

Product Code. 15515246 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 15515246

Brand: Thermo Scientific Acros 457430010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 26734-09-8
Molecular Formula C5H13NO
Molecular Weight (g/mol) 103.17
InChI Key FNVOFDGAASRDQY-UHFFFAOYSA-N
Synonym 3-amino-2,2-dimethyl-1-propanol, neopentanolamine, 1-propanol, 3-amino-2,2-dimethyl, 3-amino-2,2-dimethylpropanol, 2,2-dimethyl-3-aminopropanol, 1-amino-2,2-dimethyl-3-propanol, 3-amino-2,2-dimethyl-propan-1-ol, neo-pentanolamine, acmc-1cdms, ksc496k6d
PubChem CID 117326
IUPAC Name 3-amino-2,2-dimethylpropan-1-ol
SMILES CC(C)(CN)CO

Specifications

Melting Point 70.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 103.17
Percent Purity 97%
Chemical Name or Material 3-Amino-2,2-dimethyl-1-propanol
compliance-icons
Product Identifier
  • 3-Amino-2,2-dimethyl-1-propanol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.