missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4-bromo-1H-indazole, 97%, Thermo Scientific™

Product Code. 15466267
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15466267 250 mg 250mg
15446267 1 g 1g
15456267 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15466267 Supplier Thermo Scientific Alfa Aesar Supplier No. H33460.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Microwave heating of 2-halobenzaldehydes or 2-haloacetophenones with phenylhydrazines at 160°C for 10 min quantitatively yielded the arylhydrazones, which were further cyclized to give 1-aryl-1H-indazoles via CuI/diamine-catalyzed N-arylation under microwave heating (160°C, 10 min). Good to excellent yields were observed for 2-iodo, 2-bromo, and 2-chloro benzaldehydes or acetophenones.

Solubility
Slightly soluble.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Refrigerate 2-7°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 914311-50-5
Molecular Formula C7H6BrN3
Molecular Weight (g/mol) 212.05
MDL Number MFCD09909580
InChI Key IXXXOIXGQFPLIH-UHFFFAOYSA-N
Synonym 3-amino-4-bromo-1h-indazole, 4-bromo-1h-indazol-3-ylamine, 1h-indazol-3-amine, 4-bromo, 3-amino-4-bromoindazole, pubchem20681, acmc-209rbi, 4-bromo-1h-indazole-3-amine, 4-bromo-2h-indazol-3-ylamine, 4-bromo-1h-indazole-3-ylamine
PubChem CID 11975123
IUPAC Name 4-bromo-1H-indazol-3-amine
SMILES NC1=NNC2=CC=CC(Br)=C12

Specifications

Melting Point 142°C to 144°C
Quantity 250 mg
Solubility Information Slightly soluble.
Formula Weight 212.05
Percent Purity 97%
Chemical Name or Material 3-Amino-4-bromo-1H-indazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.