missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-6-bromopyridine, 98%, ACROS Organics™

Product Code. 10542212 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
:
1g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10542212 1g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10542212 Supplier Thermo Scientific Acros Supplier No. 360610010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 13534-97-9
CAS Min % 97.5
CAS Max % 100.0
Melting Point 75.0°C to 79.0°C
Color Beige to Black-Brown
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C5H5BrN2
MDL Number MFCD01646116
Quantity 1 g
Synonym 3-amino-6-bromopyridine, 5-amino-2-bromopyridine, 2-bromo-5-aminopyridine, 6-bromo-3-pyridinamine, 3-pyridinamine, 6-bromo, 6-bromo-pyridin-3-ylamine, 6-bromo-3-pyridylamine, 2-bromo-5-amino pyridine, 2-bromopyridin-5-amine, 6-bromopyrid-3-ylamine
InChI Key XTHKRYHULUJQHN-UHFFFAOYSA-N
SMILES C1=CC(=NC=C1N)Br
IUPAC Name 6-bromopyridin-3-amine
Molecular Weight (g/mol) 173.013
PubChem CID 642811
Formula Weight 173.01
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 3-Amino-6-bromopyridine
Show More Show Less
compliance-icons
Product Identifier
  • 3-Amino-6-bromopyridine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.