missing translation for 'onlineSavingsMsg'
Learn More

3-Aminobenzophenone, 97+%

Product Code. 11322806
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11322806 5 g 5g
11332806 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11322806 Supplier Thermo Scientific Alfa Aesar Supplier No. L08960.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 2835-78-1 | C13H11NO | 197.237 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2835-78-1
Molecular Formula C13H11NO
Molecular Weight (g/mol) 197.237
MDL Number MFCD00017103
InChI Key FUADXEJBHCKVBN-UHFFFAOYSA-N
Synonym 3-aminobenzophenone, 3-benzoylaniline, methanone, 3-aminophenyl phenyl, 3-aminophenyl phenyl methanone, benzophenone, 3-amino, m-aminobenzophenone, 3-aminophenyl phenylmethanone, 3-aminophenyl phenyl ketone, 3-amino-benzophenon, 3-amino-benzophenone
PubChem CID 17817
IUPAC Name (3-aminophenyl)-phenylmethanone
SMILES C1=CC=C(C=C1)C(=O)C2=CC(=CC=C2)N

Specifications

Melting Point 84°C to 88°C
Quantity 5 g
Beilstein 2209413
Formula Weight 197.24
Percent Purity ≥97%
Chemical Name or Material 3-Aminobenzophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.