missing translation for 'onlineSavingsMsg'
Learn More

3-(Aminomethyl)-1-methylindole, 95%, Acros Organics™

Product Code. 15284248 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15284248 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15284248 Supplier Thermo Scientific Acros Supplier No. 450490010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 19293-60-8
Infrared Spectrum Authentic
Molecular Formula C10H12N2
Quantity 1 g
Synonym 1-methyl-1h-indol-3-yl methanamine, 1-methylindol-3-yl methanamine, 1-methyl-3-indolemethylamine, 1-methyl-1h-indol-3-yl-methylamine, 1-methyl-1h-indol-3-yl methylamine, 1-1-methyl-1h-indol-3-yl methanamine, c-1-methyl-1h-indol-3-yl-methylamine, 1-1-methylindol-3-yl methanamine, n-methyl-3-aminomethyl-indol, 1-methyl-1h-indole-3-methanamine
InChI Key NOFZMDGMQKRLIV-UHFFFAOYSA-N
SMILES CN1C=C(C2=CC=CC=C21)CN
IUPAC Name (1-methylindol-3-yl)methanamine
Molecular Weight (g/mol) 160.22
PubChem CID 3934542
Formula Weight 160.22
Percent Purity ≥94.0% (HPLC)
Chemical Name or Material 3-(Aminomethyl)-1-methylindole
Show More Show Less
compliance-icons
Product Identifier
  • 3-(Aminomethyl)-1-methylindole, 95%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.