missing translation for 'onlineSavingsMsg'
Learn More

3-Aminophthalhydrazide monosodium salt, 98+%

Product Code. 11308216
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11308216 1 g 1g
11318216 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11308216 Supplier Thermo Scientific Alfa Aesar Supplier No. L15205.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 20666-12-0 | C8H9N3NaO2 | 202.169 g/mol

3-Aminophthalhydrazide monosodium salt is used for chemiluminescence analysis of metal cations, blood and glucococorticoids. It is also to image myeloperoxidase activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Aminophthalhydrazide monosodium salt is used for chemiluminescence analysis of metal cations, blood and glucococorticoids. It is also to image myeloperoxidase activity.

Solubility
Soluble in water (50 mg/ml).

Notes
Incompatible with bases, reducing agents and oxidizing agents. It is hygroscopic in nature. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20666-12-0
Molecular Formula C8H9N3NaO2
Molecular Weight (g/mol) 202.169
MDL Number MFCD00161491
InChI Key BTPIPBDXONOBET-UHFFFAOYSA-N
Synonym luminol sodium salt
PubChem CID 131673872
IUPAC Name 5-amino-2,3-dihydrophthalazine-1,4-dione;molecular hydrogen;sodium
SMILES [HH].C1=CC2=C(C(=C1)N)C(=O)NNC2=O.[Na]

Specifications

Quantity 1 g
Sensitivity Hygroscopic
Solubility Information Soluble in water (50mg/ml).
Formula Weight 199.15
Percent Purity ≥98%
Chemical Name or Material 3-Aminophthalhydrazide monosodium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.