missing translation for 'onlineSavingsMsg'
Learn More

3-Aminophthalhydrazide, Pure, SLR, Fisher Chemical™

Product Code. 11448972 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Jar
Vial
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11448972 1 g Vial 1g
10191650 5 g Glass Jar 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11448972 Supplier Fisher Chemical Supplier No. A/3150/43

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 521-31-3
Molecular Formula C8H7N3O2
MDL Number 6890

Specifications

Melting Point 319°C
Color Green to Yellow
Packaging Vial
Quantity 1 g
Formula Weight 177.16g/mol
Physical Form Solid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.