missing translation for 'onlineSavingsMsg'
Learn More

3-Aminosalicylic acid, 97%

Product Code. 11460657
Change view
Click to view available options
Quantity:
0.1 g
0.5 g
Unit Size:
0.1g
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11460657 0.5 g 0.5g
11450657 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11460657 Supplier Thermo Scientific Alfa Aesar Supplier No. B22478.02
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 570-23-0 | C7H7NO3 | 153.137 g/mol

3-Aminosalicylic acid acts as an oxygen, nitrogen donor ligand and forms complexes with silver.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Aminosalicylic acid acts as an oxygen, nitrogen donor ligand and forms complexes with silver.

Solubility
Soluble in potassium hydroxide. Slightly soluble in water.

Notes
Air and light sensitive. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 570-23-0
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.137
MDL Number MFCD00010299
InChI Key IQGMRVWUTCYCST-UHFFFAOYSA-N
Synonym 3-aminosalicylic acid, 3-amino salicylic acid, benzoic acid, 3-amino-2-hydroxy, 3-amino-2-hydroxy-benzoic acid, 2-amino-6-carboxyphenol, salicylic acid, 3-amino, 3-carboxy-2-hydroxyaniline, 3 or 5-aminosalicylic acid, benzoic acid, amino-2-hydroxy
PubChem CID 68443
IUPAC Name 3-amino-2-hydroxybenzoic acid
SMILES C1=CC(=C(C(=C1)N)O)C(=O)O

Specifications

Melting Point ∼240°C (decomposition)
Quantity 0.5 g
Beilstein 2090427
Solubility Information Soluble in potassium hydroxide. Slightly soluble in water.
Formula Weight 153.14
Percent Purity 97%
Chemical Name or Material 3-Aminosalicylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.