missing translation for 'onlineSavingsMsg'
Learn More

3-Aminosalicylic acid, MP Biomedicals™

Product Code. 11438881 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11438881 50 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11438881 Supplier MP Biomedicals Supplier No. 0215476990
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Soluble in KOH, clear solution

Specifications

CAS 570-23-0
Color Brown to Tan
Assay Percent Range 99.1% (Neutralization)
Molecular Formula C7H7NO3
Quantity 50 mg
Synonym 3-aminosalicylic acid, 3-amino salicylic acid, benzoic acid, 3-amino-2-hydroxy, 3-amino-2-hydroxy-benzoic acid, 2-amino-6-carboxyphenol, salicylic acid, 3-amino, 3-carboxy-2-hydroxyaniline, 3 or 5-aminosalicylic acid, benzoic acid, amino-2-hydroxy
InChI Key IQGMRVWUTCYCST-UHFFFAOYSA-N
SMILES C1=CC(=C(C(=C1)N)O)C(=O)O
IUPAC Name 3-amino-2-hydroxybenzoic acid
Molecular Weight (g/mol) 153.137
PubChem CID 68443
Formula Weight 153.1
Physical Form Crystalline Powder
Chemical Name or Material 3-Aminosalicylic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.