missing translation for 'onlineSavingsMsg'
Learn More

3-Aminosalicylic Acid, TRC

Product Code. 17900445
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17900445 250 mg 250mg
17922795 100 mg 100mg
17934765 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17900445 Supplier Toronto Research Chemicals Supplier No. TRCA629260250MG
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 570-23-0
Molecular Formula C7 H7 N O3
Molecular Weight (g/mol) 153.14
Synonym 3-Amino-2-hydroxybenzoic acid, Mesalazine Imp. F (EP), Benzoic acid, 3-amino-2-hydroxy-, Salicylic acid, 3-amino- (8CI), 2-Hydroxy-3-aminobenzoic acid, 3-Aminosalicylic acid, NSC 285111, TCI-A 0421
IUPAC Name 3-amino-2-hydroxybenzoic acid
SMILES Nc1cccc(C(=O)O)c1O

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C7H7NO3/c8-5-3-1-2-4(6(5)9)7(10)11/h1-3,9H,8H2,(H,10,11)
Formula Weight 153.0426
Percent Purity >95%
Chemical Name or Material 3-Amino-2-hydroxybenzoic Acid (3-Aminosalicylic Acid)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.