missing translation for 'onlineSavingsMsg'
Learn More

3-Benzyloxy-2-hydroxy-1-propylamine, 99%

Product Code. 11489370
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11489370 1 g 1g
11479370 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11489370 Supplier Thermo Scientific Alfa Aesar Supplier No. H50248.03
 Restricted Item
Request Bulk or Custom Format

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 90503-15-4 | C10H15NO2 | 181.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90503-15-4
Molecular Formula C10H15NO2
Molecular Weight (g/mol) 181.24
MDL Number MFCD00183222
InChI Key ZGHLNQMMKJNJIH-UHFFFAOYNA-N
Synonym 1-amino-3-benzyloxy-propan-2-ol, 1-amino-3-benzyloxy propan-2-ol, 2-propanol, 1-amino-3-phenylmethoxy, 1-amino-3-benzyl-oxy propan-2-ol, 3-benzyloxy-2-hydroxy-1-propylamine, 3-amino-1-phenylmethoxy propan-2-ol, 1-amino-3-benzyloxy-2-propanol, 3-benzyloxy-2-hydroxypropylamine, 1-amino-3-phenylmethoxy-propan-2-ol
PubChem CID 342795
IUPAC Name 1-amino-3-phenylmethoxypropan-2-ol
SMILES NCC(O)COCC1=CC=CC=C1

Specifications

Quantity 1 g
Formula Weight 181.24
Percent Purity 99%
Chemical Name or Material 3-Benzyloxy-2-hydroxy-1-propylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.