missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-1,2-propanediol, TRC

Product Code. 30133943
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133943 10 g 10g
17972225 5 g 5g
17911275 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133943 Supplier Toronto Research Chemicals Supplier No. TRCB68665510G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4704-77-2
Molecular Formula C3H7BrO2
Molecular Weight (g/mol) 154.99
Synonym 3-Bromo-1,2-propanediol, (±)-Glycerol 1-bromohydrin, 1-Bromo-1-deoxyglycerol, 1-Bromo-2,3-dihydroxypropane, 1-Bromo-2,3-propanediol, 2,3-Dihydroxy-1-bromopropane, 3-Bromo-1,2-dihydroxypropane, 3-Bromo-1,2-propandiol, DL-Glycerol 1-bromohydrin, Glycerol bromohydrin, Glycerol α-bromohydrin, α-Bromohydrin
IUPAC Name 3-bromopropane-1,2-diol
SMILES OCC(O)CBr

Specifications

Quantity 10 g
InChI Formula InChI=1S/C3H7BrO2/c4-1-3(6)2-5/h3,5-6H,1-2H2
Formula Weight 153.9629
Chemical Name or Material 3-Bromo-1,2-propanediol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.