missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2,6-dimethoxybenzoic acid, ≥97%, Thermo Scientific™

Product Code. 10568263
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10568263 10 g 10g
10167722 1 g 1g
10726705 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10568263 Supplier Thermo Scientific Maybridge Supplier No. BTB07593EA
 Restricted Item
Estimated Shipment: 06-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 73219-89-3
Molecular Formula C9H9BrO4
Molecular Weight (g/mol) 261.071
MDL Number MFCD00052933
InChI Key CUQANLQRQJHIQE-UHFFFAOYSA-N
PubChem CID 2774744
IUPAC Name 3-bromo-2,6-dimethoxybenzoic acid
SMILES COC1=C(C(=C(C=C1)Br)OC)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97+%
Packaging Amber glass bottle
Quantity 10 g
Synonym maybridge1_001596, acmc-1bfe7, 2,6-dimethoxy-3-bromobenzoic acid, benzoic acid,3-bromo-2,6-dimethoxy, benzoic acid, 3-bromo-2,6-dimethoxy
Formula Weight 261.07
Percent Purity 97%
Chemical Name or Material 3-Bromo-2, 6-dimethoxybenzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.