missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2,6-dimethoxybenzoic acid, 98%

Product Code. 11408546
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11408546 1 g 1g
11418546 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11408546 Supplier Thermo Scientific Alfa Aesar Supplier No. A15971.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 73219-89-3 | C9H9BrO4 | 261.071 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73219-89-3
Molecular Formula C9H9BrO4
Molecular Weight (g/mol) 261.071
MDL Number MFCD00052933
InChI Key CUQANLQRQJHIQE-UHFFFAOYSA-N
PubChem CID 2774744
IUPAC Name 3-bromo-2,6-dimethoxybenzoic acid
SMILES COC1=C(C(=C(C=C1)Br)OC)C(=O)O

Specifications

Melting Point 144°C to 148°C
Quantity 1 g
Synonym maybridge1_001596, acmc-1bfe7, 2,6-dimethoxy-3-bromobenzoic acid, benzoic acid,3-bromo-2,6-dimethoxy, benzoic acid, 3-bromo-2,6-dimethoxy
Formula Weight 261.07
Percent Purity 98%
Chemical Name or Material 3-Bromo-2,6-dimethoxybenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.