missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-fluorobenzaldehyde, 99%

Product Code. 10207080 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10207080 25 g 25g
10612572 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10207080 Supplier Thermo Scientific Acros Supplier No. 293930250

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 77771-02-9 | C7H4BrFO | 203.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77771-02-9
Molecular Formula C7H4BrFO
Molecular Weight (g/mol) 203.01
MDL Number MFCD00042186
InChI Key FAHZIKXYYRGSHF-UHFFFAOYSA-N
Synonym 4-fluoro-3-bromobenzaldehyde, benzaldehyde, 3-bromo-4-fluoro, 3-bromo-4-fluoro-benzaldehyde, 3-bromo-4-fluoro benzaldehyde, pubchem2878, acmc-209pbm, 3-bromo4fluorobenzaldehyde, 3-bromo-4fluorobenzaldehyde, 3-bromo4-fluorobenzaldehyde, 3bromo-4-fluorobenzaldehyde
PubChem CID 173604
IUPAC Name 3-bromo-4-fluorobenzaldehyde
SMILES C1=CC(=C(C=C1C=O)Br)F

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 28°C to 33°C
Color White to Yellow
Boiling Point 138°C to 139°C (2.5 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Linear Formula BrC6H3(F)CHO
Refractive Index 1.574
Quantity 25 g
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in most organic solvents
Formula Weight 203.01
Percent Purity 99%
Physical Form Low Melting Solid
Chemical Name or Material 3-Bromo-4-fluorobenzaldehyde, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Bromo-4-fluorobenzaldehyde
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.