Learn More
3'-Bromo-4'-methylacetophenone, TRC
Specifications
Specifications
| Chemical Name or Material | 3'-Bromo-4'-methylacetophenone |
| CAS | 40180-80-1 |
| Molecular Formula | C9H9BrO |
| Quantity | 2.5 g |
| Synonym | 1-(3-Bromo-4-methylphenyl)ethanone, 3-Bromo-4-methylacetophenone |
| InChI Formula | InChI=1S/C9H9BrO/c1-6-3-4-8(7(2)11)5-9(6)10/h3-5H,1-2H3 |
| SMILES | BrC1=C(C)C=CC(C(C)=O)=C1 |
| IUPAC Name | 1-(3-bromo-4-methylphenyl)ethan-1-one |
| Molecular Weight (g/mol) | 213.07 |
| Formula Weight | 211.98 |