Learn More
3-Bromo-4-methylbenzonitrile, 97%
Product Code. 11303019
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11303019 5 g 5g
11313019 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11303019 Supplier Thermo Scientific Alfa Aesar Supplier No. H27806.06
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
CAS: 42872-74-2 | C8H6BrN | 196.05 g/mol
Used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Used as pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Store in cool dry conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 42872-74-2
Molecular Formula C8H6BrN
Molecular Weight (g/mol) 196.05
MDL Number MFCD06797818
InChI Key VXUMRYMTYKDWMO-UHFFFAOYSA-N
Synonym 2-bromo-4-cyanotoluene, 4-methyl-3-bromobenzonitrile, benzonitrile, 3-bromo-4-methyl, 3-bromo-4-methylbenzenecarbonitrile, 3-bromo-4-methyl-benzonitrile, bromo p tolunitrile, bromo-p-tolunitrile, bromo-p-toluonitrile, 3-bromo-p-tolunitrile, pubchem11163
PubChem CID 14004840
IUPAC Name 3-bromo-4-methylbenzonitrile
SMILES CC1=C(Br)C=C(C=C1)C#N
Specifications
Melting Point 39°C to 43°C
Flash Point >110°C (230°F)
Quantity 5 g
UN Number UN3439
Solubility Information Insoluble in water.
Formula Weight 196.05
Percent Purity 97%
Chemical Name or Material 3-Bromo-4-methylbenzonitrile