missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-methylbenzonitrile, 97%

Product Code. 11313019
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11313019 25 g 25g
11303019 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11313019 Supplier Thermo Scientific Alfa Aesar Supplier No. H27806.14
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 42872-74-2 | C8H6BrN | 196.05 g/mol

Used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool dry conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42872-74-2
Molecular Formula C8H6BrN
Molecular Weight (g/mol) 196.05
MDL Number MFCD06797818
InChI Key VXUMRYMTYKDWMO-UHFFFAOYSA-N
Synonym 2-bromo-4-cyanotoluene, 4-methyl-3-bromobenzonitrile, benzonitrile, 3-bromo-4-methyl, 3-bromo-4-methylbenzenecarbonitrile, 3-bromo-4-methyl-benzonitrile, bromo p tolunitrile, bromo-p-tolunitrile, bromo-p-toluonitrile, 3-bromo-p-tolunitrile, pubchem11163
PubChem CID 14004840
IUPAC Name 3-bromo-4-methylbenzonitrile
SMILES CC1=C(Br)C=C(C=C1)C#N

Specifications

Melting Point 39°C to 43°C
Flash Point >110°C (230°F)
Quantity 25 g
UN Number UN3439
Solubility Information Insoluble in water.
Formula Weight 196.05
Percent Purity 97%
Chemical Name or Material 3-Bromo-4-methylbenzonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.