Learn More
3-Bromo-5-fluorobenzeneboronic acid, 95%, Thermo Scientific Chemicals
Description
3-Bromo-5-fluorobenzeneboronic acid reactant for preparation of functionalized dihalophenylboronic acid via lithiation of dihalophenyl dioxazaborocines and reaction with electrophiles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
3-Bromo-5-fluorobenzeneboronic acid reactant for preparation of functionalized dihalophenylboronic acid via lithiation of dihalophenyl dioxazaborocines and reaction with electrophiles.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
Specifications
Specifications
| Melting Point | >300°C |
| Quantity | 5 g |
| Solubility Information | Slightly soluble in water. |
| Formula Weight | 218.82 |
| Percent Purity | 95% |
| Chemical Name or Material | 3-Bromo-5-fluorobenzeneboronic acid |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.