missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-fluorobenzoic acid, 97%, Thermo Scientific™

Product Code. p-7031568
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15425447 1 g 1g
15435447 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15425447 Supplier Thermo Scientific Alfa Aesar Supplier No. H32692.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3-Bromo-5-fluorobenzoic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 176548-70-2
Molecular Formula C7H3BrFO2
Molecular Weight (g/mol) 218.00
MDL Number MFCD01569540
InChI Key KLSLJMGWUPAQGZ-UHFFFAOYSA-M
Synonym 5-bromo-3-fluorobenzoic acid, 3-fluoro-5-bromobenzoic acid, 3-bromo-5-fluorobenzoicacid, 5-fluoro-3-bromobenzoic acid, 3-bromo-5-fluoro-benzoic acid, 3-bromo-5-fluoro benzoic acid, buttpark 24\01-97, benzoic acid, 3-bromo-5-fluoro, pubchem2154, acmc-1c8tm
PubChem CID 2773339
IUPAC Name 3-bromo-5-fluorobenzoic acid
SMILES [O-]C(=O)C1=CC(F)=CC(Br)=C1

Specifications

Melting Point 141°C to 145°C
Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 219.01
Percent Purity 97%
Chemical Name or Material 3-Bromo-5-fluorobenzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.