missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Bromo-6-methoxy-2-methylpyridine, 98%

Brand:  Alfa Aesar™ H61962.06

Additional Details : CAS : 126717-59-7 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15408558

  • 67.61€ / 5g


Estimated Shipment
17-05-2021
Add to basket

Description

Description

Specifications

Chemical Identifiers

126717-59-7
202.051
VWNXCCTWVAQAPL-UHFFFAOYSA-N
10798146
CC1=C(C=CC(=N1)OC)Br
C7H8BrNO
MFCD03094943
5-bromo-2-methoxy-6-picoline, 3-bromo-6-methoxy-2-picoline, 3-bromo-6-methoxy-2-methyl-pyridine, 2-methoxy-5-bromo-6-picoline, 2-methyl-3-bromo-6-methoxypyridine, 5-bromo-2-methoxy-6-methyl pyridine, 2-methoxy-5-bromo-6-methylpyridine, 5-bromo-2-methoxy-6-methylpyridine, 5-bromo-6-methyl-2-methoxypyridine, pyridine, 3-bromo-6-methoxy-2-methyl
3-bromo-6-methoxy-2-methylpyridine

Specifications

3-Bromo-6-methoxy-2-methylpyridine
5g
C7H8BrNO
5-bromo-2-methoxy-6-picoline, 3-bromo-6-methoxy-2-picoline, 3-bromo-6-methoxy-2-methyl-pyridine, 2-methoxy-5-bromo-6-picoline, 2-methyl-3-bromo-6-methoxypyridine, 5-bromo-2-methoxy-6-methyl pyridine, 2-methoxy-5-bromo-6-methylpyridine, 5-bromo-2-methoxy-6-methylpyridine, 5-bromo-6-methyl-2-methoxypyridine, pyridine, 3-bromo-6-methoxy-2-methyl
CC1=C(C=CC(=N1)OC)Br
202.051
202.05
98%
Colorless
126717-59-7
MFCD03094943
VWNXCCTWVAQAPL-UHFFFAOYSA-N
3-bromo-6-methoxy-2-methylpyridine
10798146
Liquid
Videos
SDS Documents