Learn More
3-(Bromomethyl)-2-cyclopropyl-4-(4-fluorophenyl)quinoline, TRC
Product Code. 30133985
Change view
Click to view available options
Quantity:
100 mg
500 mg
2.5 g
Unit Size:
100mg
2.5g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133985 2.5 g 2.5g
30012332 100 mg 100mg
30009548 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30133985 Supplier Toronto Research Chemicals Supplier No. TRCB6878552.5G
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 154057-56-4
Molecular Formula C19H15BrFN
Molecular Weight (g/mol) 356.23
Synonym 2-Cyclopropyl-4-(4-fluorophenyl)-quinolyl-3-methylbromide
IUPAC Name 3-(bromomethyl)-2-cyclopropyl-4-(4-fluorophenyl)quinoline
SMILES Fc1ccc(cc1)c2c(CBr)c(nc3ccccc23)C4CC4
Specifications
Quantity 2.5 g
InChI Formula InChI=1S/C19H15BrFN/c20-11-16-18(12-7-9-14(21)10-8-12)15-3-1-2-4-17(15)22-19(16)13-5-6-13/h1-4,7-10,13H,5-6,11H2
Formula Weight 355.0372
Chemical Name or Material 3-(Bromomethyl)-2-cyclopropyl-4-(4-fluorophenyl)quinoline