missing translation for 'onlineSavingsMsg'
Learn More

3-(Bromomethyl)phenoxyacetic acid, 97%

Product Code. p-7047213
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11354885 1 g 1g
11364885 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 11354885

Brand: Thermo Scientific Alfa Aesar L18029.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 136645-25-5 | C9H9BrO3 | 245.072 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 136645-25-5
Molecular Formula C9H9BrO3
Molecular Weight (g/mol) 245.072
MDL Number MFCD02093985
InChI Key MSTODKGDFXWAIO-UHFFFAOYSA-N
Synonym 3-bromomethyl phenoxyacetic acid, 2-3-bromomethyl phenoxy acetic acid, acmc-1c8fy
PubChem CID 7010327
IUPAC Name 2-[3-(bromomethyl)phenoxy]acetic acid
SMILES C1=CC(=CC(=C1)OCC(=O)O)CBr

Specifications

Melting Point 124°C to 128°C
Quantity 1 g
UN Number UN3261
Formula Weight 245.07
Percent Purity 97%
Chemical Name or Material 3-(Bromomethyl)phenoxyacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.