missing translation for 'onlineSavingsMsg'
Learn More

3-Bromopropionyl Chloride, TRC

Product Code. 30336590
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30336590 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30336590 Supplier Toronto Research Chemicals Supplier No. TRCB67815510G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Bromopropionyl Chloride
CAS 15486-96-1
Molecular Formula C3 H4 Br Cl O
Quantity 10 g
Synonym 3-Bromopropanoyl Chloride, 3-Bromopropanoic Acid Chloride, beta-Bromopropanoyl Chloride, ω-Bromopropanoyl Chloride, beta-Bromopropionyl Chloride
InChI Formula InChI=1S/C3H4BrClO/c4-2-1-3(5)6/h1-2H2
SMILES ClC(=O)CCBr
IUPAC Name 3-bromopropanoyl chloride
Molecular Weight (g/mol) 171.42
Formula Weight 169.913
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.