missing translation for 'onlineSavingsMsg'
Learn More

3-Bromostyrene, 97%, stab. with 0.1% 4-tert-butylcatechol

Product Code. 11485957
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11485957 1 g 1g
11495957 5 g 5g
11405967 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11485957 Supplier Thermo Scientific Alfa Aesar Supplier No. B24497.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 2039-86-3 | C8H7Br | 183.048 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2039-86-3
Molecular Formula C8H7Br
Molecular Weight (g/mol) 183.048
MDL Number MFCD00000088
InChI Key KQJQPCJDKBKSLV-UHFFFAOYSA-N
Synonym 3-bromostyrene, 1-bromo-3-vinylbenzene, m-bromostyrene, benzene, 1-bromo-3-ethenyl, 1-bromo-3-vinyl-benzene, unii-6xe3sf241y, 3-bromo-styrene, pubchem23859, acmc-1cmdc, 1-bromo-3-vinylbenzene #
PubChem CID 74870
IUPAC Name 1-bromo-3-ethenylbenzene
SMILES C=CC1=CC(=CC=C1)Br

Specifications

Density 1.404
Boiling Point 73°C to 74°C (3 mmHg)
Flash Point 67°C (152°F)
Refractive Index 1.591
Quantity 1 g
Beilstein 2038490
Formula Weight 183.05
Percent Purity 97%
Chemical Name or Material 3-Bromostyrene, stabilized with 0.1% 4-tert-butylcatechol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.