missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-1-phenyl-1-propyne, 97%

Product Code. 15490408
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15490408 5 g 5g
15480408 1 g 1g
15400418 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15490408 Supplier Thermo Scientific Alfa Aesar Supplier No. H53510.06
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3355-31-5 | C9H7Cl | 150.605 g/mol

3-Chloro-1-phenyl-1-propyne is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Chloro-1-phenyl-1-propyne is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3355-31-5
Molecular Formula C9H7Cl
Molecular Weight (g/mol) 150.605
MDL Number MFCD06411085
InChI Key RMPSZEZJKSUNKR-UHFFFAOYSA-N
Synonym 3-chloroprop-1-yn-1-yl benzene, 1-phenyl-3-chloro-1-propyne, 1-phenyl-3-chloro-1-propyn, 3-chloro-1-phenyl-1-propyne, benzene, 3-chloro-1-propynyl, 3-phenylpropargyl chloride, 1-chloro-3-phenyl-2-propyne, pubchem13578, 3-chloro-l-propynyl benzene, 3-phenyl-2-propynyl chloride
PubChem CID 287343
IUPAC Name 3-chloroprop-1-ynylbenzene
SMILES C1=CC=C(C=C1)C#CCCl

Specifications

Density 1.095
Flash Point 104°C (219°F)
Linear Formula C6H4C≡CCH2Cl
Refractive Index 1.585
Quantity 5 g
Formula Weight 150.61
Percent Purity 97%
Chemical Name or Material 3-Chloro-1-phenyl-1-propyne

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.