Learn More
3-Chloro-2-fluorobenzyl bromide, 96%, Thermo Scientific™
Product Code. 11484454
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 11484454
Brand: Thermo Scientific Alfa Aesar A13114.06
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 85070-47-9
Molecular Formula C7H5BrClF
Molecular Weight (g/mol) 223.47
MDL Number MFCD00042503
InChI Key HYILLTADABKYHO-UHFFFAOYSA-N
Synonym 3-chloro-2-fluorobenzyl bromide, 1-bromomethyl-3-chloro-2-fluorobenzene, 2-fluoro-3-chlorobenzyl bromide, 3-chloro-2-fluorobenzylbromide, 3-chloro-2-fluoro benzyl bromide, alpha-bromo-3-chloro-2-fluorotoluene, alpha-bromo-2-fluoro-3-chlorotoluene, benzene, 1-bromomethyl-3-chloro-2-fluoro, 1-bromomethyl-3-chloro-2-fluoro-benzene
PubChem CID 522827
IUPAC Name 1-(bromomethyl)-3-chloro-2-fluorobenzene
SMILES FC1=C(Cl)C=CC=C1CBr
Specifications
Melting Point 36°C to 40°C
Boiling Point 125°C to 126°C (15 mmHg)
Refractive Index 1.569
Quantity 5 g
UN Number UN3261
Formula Weight 223.48
Percent Purity 96%
Chemical Name or Material 3-Chloro-2-fluorobenzyl bromide