missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-2-methylbenzeneboronic acid, 97%

Product Code. 15453997
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15453997 250 mg 250mg
15443997 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15453997 Supplier Thermo Scientific Alfa Aesar Supplier No. H30893.MD
 Restricted Item
Estimated Shipment: 02-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 313545-20-9 | C7H8BClO2 | 170.399 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 313545-20-9
Molecular Formula C7H8BClO2
Molecular Weight (g/mol) 170.399
MDL Number MFCD04115641
InChI Key BJPNVVXTUYMJPN-UHFFFAOYSA-N
Synonym 3-chloro-2-methylphenyl boronic acid, 2-borono-6-chlorotoluene, 3-chloro-2-methylbenzeneboronic acid, 3-chloro-2-methylphenyboronic acid, boronic acid, 3-chloro-2-methylphenyl, boronic acid,b-3-chloro-2-methylphenyl, 3-chloro-2-methyl-phenyl boronic acid, acmc-209hls, 3-chloro-2-methylphenboronic acid, 3-chloro-2-methyl-phenylboronic acid
PubChem CID 3744103
IUPAC Name (3-chloro-2-methylphenyl)boronic acid
SMILES B(C1=C(C(=CC=C1)Cl)C)(O)O

Specifications

Quantity 250 mg
Formula Weight 170.4
Percent Purity 97%
Chemical Name or Material 3-Chloro-2-methylbenzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.