missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-fluoro-5-nitrobenzotrifluoride, 98%, Thermo Scientific Chemicals

Product Code. 11304246
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11304246 5 g 5g
11394236 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11304246 Supplier Thermo Scientific Alfa Aesar Supplier No. L19322.06
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 101646-02-0 | Molecular Formula: C7H2ClF4NO2 | Molecular Weight: 243.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101646-02-0
Molecular Formula C7H2ClF4NO2
Molecular Weight (g/mol) 243.54
MDL Number MFCD00674108
InChI Key JVOKKJHIEVEFEB-UHFFFAOYSA-N
Synonym 3-chloro-4-fluoro-5-nitrobenzotrifluoride, 1-chloro-2-fluoro-3-nitro-5-trifluoromethyl benzene, 3-chloro-2-fluoro-1-nitro-5-trifluoromethyl benzene, 5-chloro-4-fluoro-3-nitrotrifluoromethylbenzene, 5-chloro-4-fluoro-3-nitro-trifluoromethylbenzene
PubChem CID 2736565
IUPAC Name 1-chloro-2-fluoro-3-nitro-5-(trifluoromethyl)benzene
SMILES [O-][N+](=O)C1=C(F)C(Cl)=CC(=C1)C(F)(F)F

Specifications

Density 1.61
Boiling Point 200°C
Flash Point >110°C (230°F)
Refractive Index 1.482
Quantity 5 g
UN Number UN2810
Beilstein 5436538
Formula Weight 243.55
Percent Purity 98%
Chemical Name or Material 3-Chloro-4-fluoro-5-nitrobenzotrifluoride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.