missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-5-fluorobenzeneboronic acid, 95%

Product Code. 15424827
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15424827 1 g 1g
15434827 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15424827 Supplier Thermo Scientific Alfa Aesar Supplier No. H32137.03
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 328956-61-2 | C6H5BClFO2 | 174.36 g/mol

3-Chloro-5-fluorobenzeneboronic acid is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Chloro-5-fluorobenzeneboronic acid is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 328956-61-2
Molecular Formula C6H5BClFO2
Molecular Weight (g/mol) 174.36
MDL Number MFCD06801741
InChI Key XYQDHVBKUNNLGZ-UHFFFAOYSA-N
Synonym 3-chloro-5-fluorobenzeneboronic acid, 3-chloro-5-fluorophenyl boronic acid, boronic acid, 3-chloro-5-fluorophenyl, pubchem6363, acmc-1agjy, ksc497k5n, 3-borono-5-fluorochlorobenzene, 3-chloro-5-fluoro-benzeneboronic acid, 3-chloro-5-fluoro-phenyl boronic acid, 3-chloranyl-5-fluoranyl-phenyl boronic acid
PubChem CID 15391287
IUPAC Name (3-chloro-5-fluorophenyl)boronic acid
SMILES OB(O)C1=CC(F)=CC(Cl)=C1

Specifications

Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 174.37
Percent Purity 95%
Chemical Name or Material 3-Chloro-5-fluorobenzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.