Learn More
3-Chloro-5-(trifluoromethoxy)phenol, 97%, Thermo Scientific™
Product Code. 15475327
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. 15475327
Brand: Thermo Scientific Alfa Aesar H32579.MD
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1017778-52-7
Molecular Formula C7H4ClF3O2
Molecular Weight (g/mol) 212.55
MDL Number MFCD09832330
InChI Key GDQVMGRMDBOXLX-UHFFFAOYSA-N
Synonym 3-chloro-5-trifluoromethoxy phenol, 5-chloro-3-trifluoromethoxy phenol, 3-chloro-5-trifluoromethoxy phenol, jrd
PubChem CID 46737653
IUPAC Name 3-chloro-5-(trifluoromethoxy)phenol
SMILES OC1=CC(OC(F)(F)F)=CC(Cl)=C1
Specifications
Refractive Index 1.4702
Quantity 250 mg
Formula Weight 212.56
Percent Purity 97%
Chemical Name or Material 3-Chloro-5-(trifluoromethoxy)phenol