Learn More
3-Chloro-N-methylpropan-1-amine Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | 3-Chloro-N-methylpropan-1-amine Hydrochloride |
| CAS | 97145-88-5 |
| Molecular Formula | C4H10ClN•(HCl) |
| Quantity | 250 mg |
| Synonym | Fluoxetine Impurity, 3-(Methylamino)propyl Chloride Hydrochloride, N-Methyl(3-chloropropyl)amine Hydrochloride |
| InChI Formula | InChI=1S/C4H10ClN.ClH/c1-6-4-2-3-5;/h6H,2-4H2,1H3;1H |
| SMILES | CNCCCCl.Cl |
| IUPAC Name | 3-chloro-N-methylpropan-1-amine;hydrochloride |
| Molecular Weight (g/mol) | 107.583646 |
| Formula Weight | 143.0269 |