missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorobenzenamine, TRC

Product Code. 30134915
Change view
Click to view available options
Quantity:
10 g
100 g
Unit Size:
100g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134915 100 g 100g
30005272 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30134915 Supplier Toronto Research Chemicals Supplier No. TRCC364610100G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-42-9
Molecular Formula C6 H6 Cl N
Molecular Weight (g/mol) 127.57
Synonym Aniline, m-chloro- (8CI), 3-Chlorobenzenamine, 1-Amino-3-chlorobenzene, 3-Amino-1-chlorobenzene, 3-Chloro-1-aminobenzene, 3-Chloroaniline, 3-Chlorophenylamine, 5-Chloroaniline, Fast Orange GC Base, NSC 17581, Orange GC base, m-Aminochlorobenzene, m-Chloroaniline, m-Chlorophenylamine
IUPAC Name 3-chloroaniline
SMILES Nc1cccc(Cl)c1

Specifications

Quantity 100 g
InChI Formula InChI=1S/C6H6ClN/c7-5-2-1-3-6(8)4-5/h1-4H,8H2
Formula Weight 127.0189
Chemical Name or Material 3-Chloroaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.