missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorobenzyl Bromide 98+%, ACROS Organics™

Product Code. 10275630 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 mL
Packaging:
Glass bottle
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10275630 50 mL Glass bottle 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10275630 Supplier Thermo Scientific Acros Supplier No. 159640500

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 766-80-3
CAS Min % 98.0
CAS Max % 100.0
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Molecular Formula C7H6BrCl
Refractive Index 1.5870 to 1.5890 (20°C, 589nm)
MDL Number MFCD00000597
Quantity 50 mL
Synonym 3-chlorobenzyl bromide, 1-bromomethyl-3-chlorobenzene, 3-chlorobenzylbromide, alpha-bromo-3-chlorotoluene, m-chlorobenzyl bromide, benzene, 1-bromomethyl-3-chloro, alpha-bromo-m-chlorotoluene, .alpha.-bromo-m-chlorotoluene, 1-bromomethyl-3-chloro-benzene, 3-chorobenzylbromide
InChI Key LZIYAIRGDHSVED-UHFFFAOYSA-N
SMILES ClC1=CC=CC(CBr)=C1
IUPAC Name 1-(bromomethyl)-3-chlorobenzene
Molecular Weight (g/mol) 205.48
PubChem CID 69838
Formula Weight 205.48
Percent Purity ≥98.0%
Physical Form Liquid
Chemical Name or Material 3-Chlorobenzyl bromide, 98+%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.